Anti-MUC1 Recombinant Antibody [Clone: Mc5]
Catalog # 76630-242
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:HGNC:HGNC:7508
- Antigen Symbol:MUC1
- Clonality:Recombinant
- Clone:Mc5
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:4582
- Antigen Synonyms:Episialin|CA 15-3|PEMT|PUM|Peanut-reactive urinary mucin|CD227|Mucin-1|KL-6|BEM|EMA|Carcinoma-associated mucin|Krebs von den Lungen-6|Cancer antigen 15-3|PEM|H23AG|Breast epithelial mucin|Breast carcinoma-associated antigen DF3|Tumor-associated epithelial membrane antigen|Tumor-associated mucin|Polymorphic epithelial mucin
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Human breast epithelial mucin.
- Tested Applications:IRA
- Cat. No.:76630-242
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HGNC:HGNC:7508
Clonality: Recombinant
Clone: Mc5
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human