Anti-RHO Recombinant Antibody [Clone: Rho 1D4]
Catalog # 76630-562
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:rhodopsin
- Antigen Symbol:RHO
- Clonality:Recombinant
- Clone:Rho 1D4
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Zebrafish,Amphibian,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:6010
- Antigen Synonyms:Rho-tag|Visual Purple
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Bleached bovine rod outer segment (ROS).
- Tested Applications:Radioimmune labeling
- Cat. No.:76630-562
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: rhodopsin
Clonality: Recombinant
Clone: Rho 1D4
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Mouse;Rat;Bovine;Human;Zebrafish;Amphibian