Anti-CCR3 Recombinant Antibody [Clone: 5E8-G9-B4]
76630-150
: Prosci
- Antibody Type:Primary
- Antigen Name:C-C motif chemokine receptor 3
- Antigen Symbol:CCR3
- Clonality:Recombinant
- Clone:5E8-G9-B4
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:1232
- Antigen Synonyms:C-C Motif Chemokine Receptor 3
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Cat. No.:76630-150
- Packaging:Vial
Type: Primary
Antigen: C-C motif chemokine receptor 3
Clonality: Recombinant
Clone: 5E8-G9-B4
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human