Anti-AOC3 Recombinant Antibody [Clone: TK8-14]
Catalog # 76630-240
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:amine oxidase copper containing 3
- Antigen Symbol:AOC3
- Clonality:Recombinant
- Clone:TK8-14
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:8639
- Antigen Synonyms:Amine oxidase|copper containing 3|Vascular adhesion protein 1|Membrane primary amine oxidase|Semicarbazide-sensitive|HPAO|copper-containing amine:oxy|SSAO|AOC3|VAP1|Copper amine oxidase|amine oxidase
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Affinity purified VAP-1 from tonsil stroma of human origin.
- Cat. No.:76630-240
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: amine oxidase copper containing 3
Clonality: Recombinant
Clone: TK8-14
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human