NEBNext® FFPE DNA Library Preparation Kits
Supplier: New England Biolabs (NEB)
Certificates
About this item
This kit is designed for use with pre-sheared FFPE DNA.
- Includes FFPE DNA repair reagents plus optimized library prep reagents and protocol
- Increased library yields
- Improved sequencing metrics
- Greater sensitivity of somatic variant calling
- Automation-friendly workflow
The NEBNext FFPE DNA Library Prep Kit includes the NEBNext FFPE DNA Repair v2 Mix, an optimized cocktail of enzymes designed to repair FFPE DNA, library prep reagents featuring a new polymerase master mix, and a protocol optimized for FFPE DNA. This kit is designed for use with pre-sheared FFPE DNA.
Frequently Bought Together