Order Entry
United States
ContactUsLinkComponent
CardioChek Plus Lipid Panel Sarter Kit With Printer
CardioChek Plus Lipid Panel Sarter Kit With Printer
Catalog # 77148-112
Supplier:  PTS Diagnostics
CAS Number:  
undefined
CardioChek Plus Lipid Panel Sarter Kit With Printer
Catalog # 77148-112
Supplier:  PTS Diagnostics
Supplier Number:  PTS906
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    CardioChek Plus lipid panel with printer starter kit
  • Operating Temperature Range:
    20...30 °C (68...86 °F)
  • Storage Temperature:
    10...40 °C (50...104 °F)
  • Cat. No.:
    77148-112

Specifications

About this item

CardioChek Plus lipid panel starter kit with printer provides full lipid panel test results in as little as 90 seconds at the point of care.

Certifications: CLIA-waived, FDA-cleared, Meets NCEP guidelines for accuracy and precision, CRMLN-certified for CHOL and HDL tests.
Ordering information: CardioChek Plus lipid panel with printer starter kit includes: (1) CardioChek Plus analyzer, (4) PTS panels lipid panels (60 lipid tests), (1) PTS panels multi-chemistry controls, (4) PTS collect capillary tubes, 40 µl (64 capillary tubes total), (1) CardioChek printer (includes 1 roll of labels).