Order Entry
United States
ContactUsLinkComponent
CardioChek Plus Smart Bundle Starter Kit with Printer
CardioChek Plus Smart Bundle Starter Kit with Printer
Catalog # 77148-108
Supplier:  PTS Diagnostics
CAS Number:  
undefined
CardioChek Plus Smart Bundle Starter Kit with Printer
Catalog # 77148-108
Supplier:  PTS Diagnostics
Supplier Number:  PTS761
CAS Number:  

Specifications

  • Description:
    CardioChek Plus smart bundle with printer starter kit
  • Operating Temperature Range:
    20...30 °C (68...86 °F)
  • Storage Temperature:
    10...40 °C (50...104 °F)
  • Cat. No.:
    77148-108

Specifications

About this item

CardioChek Plus smart bundle starter kit with printer provides full lipid panel and glucose test results in as little as 90 seconds at the point of care.

Certifications: CLIA-waived, FDA-cleared, Meets NCEP guidelines for accuracy and precision, CRMLN-certified for CHOL and HDL tests.
Ordering information: CardioChek Plus smart bundle with printer starter kit includes: (1) CardioChek Plus analyzer, (4) PTS panels Lipid + eGlu smart bundles (60 lipid + glucose tests), (1) PTS panels multi-chemistry controls, (4) PTS collect capillary tubes, 40 µl (64 capillary tubes total), (1) CardioChek printer (includes 1 roll of labels).