CardioChek Plus Smart Bundle Starter Kit
Catalog # 77148-106
Supplier: PTS Diagnostics
CAS Number:
Specifications
- Description:CardioChek Plus smart bundle starter kit
- Operating Temperature Range:20...30 °C (68...86 °F)
- Storage Temperature:10...40 °C (50...104 °F)
- Cat. No.:77148-106
Specifications
About this item
CardioChek Plus smart bundle starter kit provides full lipid panel and glucose test results in as little as 90 seconds at the point of care.
Certifications: CLIA-waived, FDA-cleared, Meets NCEP guidelines for accuracy and precision, CRMLN-certified for CHOL and HDL tests.
Ordering information: CardioChek Plus smart bundle starter kit includes: (1) CardioChek Plus analyzer, (4) PTS panels lipid + eGlu smart bundles (60 lipid + glucose tests), (1) PTS panels multi-chemistry controls, (4) PTS collect capillary tubes, 40 µl (64 capillary tubes total).