Eisco A-Shaped Retort Bases with Leveling Screws
Supplier: Wards
Certificates
About this item
A-Shaped Base with leveling screws for optimized stability made of chemically-resistant cast iron.
- A-shaped retort bases made of chemical-resistant cast iron with a black coating
- Leveling screws on two ends for achieving precise and stable positioning
- Heavy and sturdy construction provides superb stability for experimental setups
- Features a centered-tapped hole for a 10×1.5 mm threaded rod (not included)
Heavy, A-shaped Retort Stands are made of chemically-resistant cast iron with a black finish. Features leveling screws on two ends for achieving precise and stable positioning, and a tapped hole at the center of the crossbar, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.
Frequently Bought Together