Liquid Batch Sampling System, LS-20
Catalog # 77093-404
Supplier: Lighthouse Worldwide
Specifications
- Description:Liquid batch sampling system LS-20
- Cat. No.:77093-404
Specifications
About this item
Lighthouse’s Liquid Batch Samplers are designed to be user-friendly and intuitive for utilization. Most importantly, it is designed to be compliant to industry standards.
- 1.0 to 50 µm or 1.0 to 200 µm size range
- Integrated 'light obscuration' particle counter software designed for 21 CFR part 11
- International regulatory EP, JP reports
- USP 788 reports
- Sample volumes from 1 to 1000 ml
- Preconfigured USP 788 recipes
- Precise syringe pump flow control
- Stainless steel enclosure
- Built-in magnetic stirrer
- Automated sample handling maximizes repeatability
- Compact footprint