Specifications
- Description:Scan air pro scanner
- Cat. No.:77093-410
Specifications
About this item
The Lighthouse Scan Air provides an easy-to-use means of checking HEPA/ULPA filters and their seals for leaks and efficiency.
- Ergonomic and lightweight
- Touchscreen user interface
- Upstream and downstream measurement modes
- Automatic alarm threshold calculation
- Display percentage leak during downstream mode
- Only solution for testing PTFE media filters
- Ability to challenge test filters with a much lower concentration than a photometer
- Virtually no coalescence of aerosol
- Use PSL and PAO for filter challenge testing
- Multi-function system allows instruments to be utilized for other applications
The Scan air incorporates an isokinetic probe specifically designed for sampling a flow rate of 1.0 CFM (28.3 LPM) across a wide area. The unit includes a remote start/stop button and 7.62 m (25") cable. The remote switch features hands-free operation of the Solair portable particle counter, enabling the user to efficiently sample in compliance with ISO 14644.
The on-board alarm buzzer and LED quickly notify the user if a count exceeds the alarm threshold, even when the Scan Air is being used in a high noise environment. The twist-lock connector quickly and easily connects or disconnects the Scan Air from a Solair portable particle counter.
The Solair and Scan Air can be used as a portable contamination detection system throughout the cleanroom environment.