Liquid Grab Sampler
Product Details & Documents
The Liquid Grab Sampler (LGS) adds value to the Lighthouse remote liquid particle counter by providing a reliable pump system and NEMA enclosure to help integrate it with process equipment and large facility monitoring systems.
- External pump control
- RS-485 Modbus output
- Easy to use features
- Building management system interface
- Maintains high flow rate: 100 ml/min (user controllable)
- Same robust data features as the Lighthouse remote liquid particle counter
- Designed for reliability
- Easily download data to computer
The Liquid Grab Sampler (LGS) was designed to provide reliable sampling in a variety of applications such as parts washing, waste water monitoring, and process monitoring for many different environments. It incorporates a state-of-the-art pump, precision pump speed control, precision flow meter, and externally controlled low voltage pump switching. The LGS is easy to configure, operate, and monitor. The Remote Liquid Particle Counter (sold separately) provides particle count data in real time. All data can be transferred to a single computer or a facility monitoring system.
Specifications
- Description:Liquid grab sampler