Order Entry
United States
ContactUsLinkComponent
Circular Specimen Chuck 30 mm Kit for Leica, TBS, And Tanner Cryostats
Circular Specimen Chuck 30 mm Kit for Leica, TBS, And Tanner Cryostats
Catalog # 76948-578
undefined
Circular Specimen Chuck 30 mm Kit for Leica, TBS, And Tanner Cryostats
Catalog # 76948-578
Supplier Number:  SP103-KT

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Type:
    Circular chuck style
  • Description:
    Specimen chuck kit
  • Cat. No.:
    76948-578

Specifications

About this item

SP103-KT: (2) 25 mm, (6) 30 mm, and (2) 40 mm in different colors, an angular brush, and a teasing needle.

  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats
Delivery: Standard kit includes 10 common Leica-type specimen chucks, an angular brush and a teasing needle.