ContactUsLinkComponent
Nilfisk Nommo Foam/Rinse/Sanitize Unit
Nilfisk Nommo Foam/Rinse/Sanitize Unit
Catalog # 77007-824
Supplier:  Unimed-Midwest
undefined
Nilfisk Nommo Foam/Rinse/Sanitize Unit
Catalog # 77007-824
Supplier:  Unimed-Midwest
Supplier Number:  FFRS516MMO
Specifications
  • Description:
    Nilfisk nommo foam/rinse/sanitize unit
  • Cat. No.:
    77007-824
Specifications
About this item
Clean and sanitize small to medium spaces with the water-driven Nommo. Three functions in one unit save space and simplify the sanitation process.
  • Powered by water
  • Draws from two concentrated products, one at a time
  • Color-coded selector valve matches chemical suction lines
  • Control dilution rate for each product independently using metering tips
  • Spray or foam release; switch function by swapping nozzles
  • Simple installation and operation
The unit is powered by water, requiring 29 to 87 psi (2 to 6 bar) of incoming water pressure, and draws from two concentrated products, one at a time.
Choose from two nozzle types; the patented Tornado rinse/spray nozzle provides high-impact output with 20% less water, while the top jet foam nozzle creates visible, wet foam without compressed air.