Oakton Replacement Turbidity Calibration Set, Antylia Scientific
76795-394
:
- Description:Replacement Turbidity Calibration Solution Kit; 0.02, 20, 100, and 800 NTU Standards
- Size:4 x 60 ml
- Cat. No.:76795-394
- For use with:Oakton T-100 Turbidity Meter
For calibration of Oakton T-100 turbidity meters
- Includes one each of 60-mL HPDE bottle of four NTU standards