Science Olympiad 2024 Air Trajectory Kit
Catalog # 470352-140
Supplier: Avantor
Specifications
- Description:2024 air trajectory kit
- No. of lab groups:1
- Cat. No.:470352-140
Specifications
About this item
Prepare for upcoming competitions with this practice kit, the only kits officially licensed and approved by science olympiad.
- Designed hand-in-hand with the national event supervisors
- Aligned to NGSS-based science olympiad events
- 1 launcher can be built from this kit
- Event rules can be downloaded FREE from the science olympiad website: soinc.org/rules-2024
This kit features components hand selected by the science olympiad organization.
The 2023 to 2024 science olympiad kits were developed to provide an introduction to each event and are not intended to be an 'out of the box solution' for competition. Students and competitors should practice innovation and creativity to make their design their own competition-ready device/activity.
Ordering information: Kit contains: (2) 67 cm pieces 1" schedule 40 PVC, (2) 30 cm pieces 1" schedule 40 PVC, (2) 20 cm pieces 1" schedule 40 PVC, (2) 17 cm pieces 1" schedule 40 PVC, (2) 15 cm pieces 1" schedule 40 PVC, 25 cm piece 1.5" schedule 40 PVC, (6) 1" schedule 40 PVC 90 degree elbow, (2) 1" schedule 40 PVC T-joints, 1.5" PVC end cap, 1/4-20 threaded rod (12"), (6) ¹/₄ to 20 nuts, (2) nylon hose barbs (³/₈ ID × ¹/₈ MIP), and 12" plastic tubing.