ContactUsLinkComponent
PSA™ iFluor® 594 Imaging Kit with Goat Anti-Rabbit IgG
PSA™ iFluor® 594 Imaging Kit with Goat Anti-Rabbit IgG
Catalog # 76484-920
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
PSA™ iFluor® 594 Imaging Kit with Goat Anti-Rabbit IgG
Catalog # 76484-920
Supplier:  AAT BIOQUEST INC
Supplier Number:  45230
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 - 50 times higher than the widely used tyramide (TSA) reagents
  • Size:
    100 tests
  • Cat. No.:
    76484-920
Specifications
About this item
iFluor® 594 PSA kit is a much superior replacement for Alexa Fluor 594 tyramide-based kit or other spectrally similar fluorescent tyramide or TSA kits.
  • Higher efficiency
  • Improved fluorescence signal
  • Less consumption of primary antibody
  • 10 to 50X improvement over TSA reagents
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.
Packaging: Plastic vial.
Caution: For research use only.