VirusGEN® AAV Transfection Kits with RevIT™ AAV Enhancer
Supplier: Mirus Bio
Certificates
About this item
This novel enhancer is easily integrated into existing workflows, and it produces 2-4x higher titers across a range of serotypes, cellular growth media formulations, and transfection platforms. RevIT™ AAV Enhancer can further drive down upstream manufacturing costs by permitting the use of lower levels of plasmid DNA while maintaining high-quality titers. Continual improvements in both upstream and downstream AAV manufacturing processes will allow the next generation of gene therapies to reach a broader audience
- Designed to deliver a higher titer for any serotype, reagent, media, or cell line.
- 2-4X increase in AAV titer.
- >50% reduction in cost per dose.
- 2X doses produced per bioreactor run.
Specifications
- Bioproduction:Yes
- Cell Specific:HEK 293
- DNA:Yes
- Genome editing:Yes
- Virus production:Yes
- Storage Temperature:−20 °C
Specifications
Frequently Bought Together