TransIT-VirusGEN® Transfection Reagents
TransIT-VirusGEN® Transfection reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant Adeno-Associated virus (AAV) and Lentivirus production.
- Performance – Efficient DNA delivery for production of high-titer viral vectors
- Scalability – Efficient production from <1 ml to large scale cultures
- Flexibility – Compatible with different recombinant virus and cell culture systems
- Animal origin free – Fully synthetic transfection reagent formulation
TransIT-VirusGEN® is ideal for academic research and biopharmaceutical discovery. Titers can be further increased with optimized enhancers included in the VirusGEN® AAV Transfection Kit and VirusGEN® LV Transfection kit.
- Bioproduction:Yes
- DNA:Yes
- Genome editing:Yes
- Protein Delivery:Yes
- Virus production:Yes
Frequently Bought Together