ReadiLink™ iFluor® 555 FISH Fluorescence Imaging Kit
76779-084
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
- Description:Convenient tool for labeling a target DNA using an iFluor® 555 labeled FISH probe via in situ hybridization, for PCR microplates
- Size:25 reactions
- Cat. No.:76779-084
The kit provides Taq DNA polymerase enzyme which incorporates iFluor® 555-dUTPs in the target DNA through Polymerase Chain Reaction (PCR).
- Robust protocol
- Bright labeling
- Photostable fluorescence
The Readilink™ iFluor® 555 FISH fluorescence imaging kit is a convenient tool for labeling a target DNA using an iFluor® 555 labeled FISH probe via in situ hybridization.
Our proprietary iFluor® dyes are brighter and more photostable than traditional fluorescent labels, providing the desired resolution and signal.
: Packaged in plastic vials.
: For research use only.