PIP® Bouton® Universal Fit Polycarbonate Safety Visor
Catalog # 76944-632
Supplier: PIP
Specifications
- Description:Universal fit safety visor
- Material:Polycarbonate
- Thickness:0.040"
- Cat. No.:76944-632
Specifications
About this item
Universal fit polycarbonate safety visor with 0.040" thick lens.
- Made with virgin resin for better uniformity of the viewing area
- Universal hole pattern will fit most headgear and hard hat adapters
- Ships with masking on both sides to prevent scratching
Polycarbonate safety visor with aluminum binding is ideal for high impact applications. Outperforms PETG (Polyethylene Terephthalate Glycol) in a wider range of temperatures.
Certifications: Meets ANSI Z87.1. Meets CSA Z94.3.