Label IT® MFP488 Nucleic Acid Labeling Kits
Supplier: Mirus Bio
Certificates
About this item
Nucleic acid labeling reagents; label any DNA or RNA template – suitable for a range of applications.
- High photostability – facilitates longer time for observation and image capture
- Widespread Instrument compatibility – spectrally similar to fluorescein
- Stable at Low pH – fluorescence is maintained over a broad range (pH 4 to 9)
- One-step chemical method – easily and consistently control the labeling reactions
- Adjustable labeling density – Achieve high sensitivity with optimally labeled DNA and RNA
- Covalent mechanism – permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance
NOTE: All the Label IT® reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
Specifications
- DNA and siRNA:Yes
- DNA:Yes
- dsRNA:Yes
Specifications
Related Information
Mirus Bio DNA & RNA labelling, In vivo Delivery and Companion Products
Product Literature
Frequently Bought Together