ContactUsLinkComponent
Wards® Exothermic/Endothermic Combination Demonstration
Wards® Exothermic/Endothermic Combination Demonstration
Catalog # 470337-924
Supplier:  Ward's Science
undefined
Wards® Exothermic/Endothermic Combination Demonstration
Catalog # 470337-924
Supplier:  Ward's Science
Supplier Number:  IS7006
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Exothermic/endothermic combination demonstration
  • Cat. No.:
    470337-924
  • Includes:
    SDS and instructions
  • No. of uses:
    5
Specifications
About this item
Study endothermic and exothermic reactions with this hands-on demonstration.
  • Contains enough materials for 5 demonstrations
Study both processes of endothermic and exothermic reactions. Reactions are safely carried out in plastic bags. The students can pass the bags around to feel the results of the reactions.
Delivery: 5 bags of the following mixture: 25 g Iron metal powder; 1 g Sodium chloride crystals; 5 g Calcium chloride flakes; 2 tbsp Vermiculite; 8 package×2 tablets Sodium bicarbonate/Citric acid tablets; 15 Plastic cups; 15 Stirring sticks; 15 Bags containing: 25 g Ammonium nitrate, 1 tbsp Vermiculite.