- Description:Formation of tin wool demo
- Cat. No.:470337-912
- Includes:SDS and instructions
- No. of uses:7
Form tin wool through a single replacement reaction.
- Includes enough materials for 7 demonstrations
This is a single replacement reaction in which the zinc metal dissolves in solution and tin metal is formed.