ContactUsLinkComponent
Wards® Solid Fuel Chemical Demonstration Kit
Wards® Solid Fuel Chemical Demonstration Kit
Catalog # 470337-906
Supplier:  Ward's Science
undefined
Wards® Solid Fuel Chemical Demonstration Kit
Catalog # 470337-906
Supplier:  Ward's Science
Supplier Number:  IS7025
Specifications
  • Description:
    Solid fuel chemical demonstration kit
  • Cat. No.:
    470337-906
  • Includes:
    SDS and instructions
  • No. of uses:
    2
Specifications
About this item
Explore a solid that can be ignited with a match!
  • Contains enough materials for 2 demonstrations
Two solutions are poured together in a beaker and form a gel. The gel can be ignited with a match. Commercially this gel is sold as Sterno®.
Delivery: Includes 2×25 ml Ethyl alcohol, denatured; 2×25 ml Calcium acetate solution.