Estes Preassembled Rocketry Parachutes
Supplier: Estes Industries
Certificates
About this item
Lightweight, tear-resistant parachutes in a variety of sizes.
- Blown film plastic is lightweight yet durable
- Hexagonal shape, with shroud lines pre-attached in three loops
- Shroud lines will not break in flight, nor fray over time
- Multiple sizes for any sized rocket
These parachutes come preassembled and ready to use on your choice of model rockets. The shroud lines come attached, and it simply attaches to the nose cone by sliding the shroud lines through the anchor then looping the parachute through them. This lightweight but strong parachute is designed to fit into small airframes and bring the rocket down to the ground with a light touch. Perfect for hobbyists, or use them in the science classroom to study the effects of drag as a function of diameter!
Frequently Bought Together