ContactUsLinkComponent
Estes Alpha III Launch Set
Estes Alpha III Launch Set
  470351-942
undefined
Estes Alpha III Launch Set
  470351-942
 :  1427
  • Description:
    Alpha III launch set
  • Cat. No.:
    470351-942
The iconic Alpha III model rocket, complete with launch pad and controller.
  • Beginner-level build
  • Recommended engines: A8-3, B4-4, B6-4
  • Max. altitude 351 m (1150')
  • Launch system included
Ready to enter the exciting world of model rocketry? The Alpha III launch set is a perfect kit to get started, with almost everything you'll need for your first launch. The Alpha III was the first rocket in the beginner level series. With easy-to-assemble parts, you’ll be ready for liftoff in an hour: perfect for teachers who want to include rocketry in their classroom. The included Porta pad II launch system will have you and your new rocket out in the field in no time!
 : Required but not included: (4) AA batteries, rocket engines.
 : Kit includes (1) Alpha III model rocket with parachute recovery, (1) Porta pad II, and (1) electron beam launch controller.