Order Entry
United States
ContactUsLinkComponent
7-Fluoro-2,3-dihydropyrrolo[2,1-b]quinazolin-9(1H)-one 95%
7-Fluoro-2,3-dihydropyrrolo[2,1-b]quinazolin-9(1H)-one 95%
Catalog # 77104-120
Supplier:  Ambeed
CAS Number:  
7-Fluoro-2,3-dihydropyrrolo[2,1-b]quinazolin-9(1H)-one 95%
Catalog # 77104-120
Supplier:  Ambeed
Supplier Number:  A608276-1G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    77104-120

Specifications

About this item

 :
 :204.20 g/mol
 :GHS07
CAS Number:380638-36-8
Formula:C₁₁H₉FN₂O
MDL Number:MFCD02333781