ContactUsLinkComponent
SCIEX Triple Quad 5500+ LC-MS/MS Systems
SCIEX Triple Quad 5500+ LC-MS/MS Systems
Supplier:  SCIEX
Certificates
undefined
SCIEX Triple Quad 5500+ LC-MS/MS Systems
Supplier:  SCIEX
Specifications
  • Catalog No:
  • 76644-994
  • 76644-990
  • Model:
  • Triple Quad 5500+ system, QTRAP activated mass spectrometer
  • Triple Quad 5500+ system mass spectrometer (QTRAP ready)
  • Detector:
  • HED detector and high capacity CEM (Increased productivity with fast polarity switching of 5 ms enabling up to 6 orders of magnitude of linear dynamic range)
  • HED detector and high capacity CEM (Increased productivity with fast polarity switching of 5 ms enabling up to 6 orders of magnitude of linear dynamic range)
  • Mass range:
  • 50 - 1000, QTRAP (dynamic range 6 orders of magnitude)
  • 5 - 1250, Triple Quad
  • Polarity switching:
  • 5 ms
  • 5 ms
  • Scan:
  • Enhanced MS scan, enhanced product ion scan, enhanced resolution scan, MS3 scan, MRM3 scan and TripleTrap scanning modes
  • Full scan MS and selected ion for both Q1 and Q3, product ion scan, precursor ion scan, neutral loss or gain scan, multiple reaction monitoring (MRM)
  • Scan speed:
  • 20000 Da/s max.
  • 12000 Da/s max.