OPTI-LYNX™ In-Line Guard Holder Kit
Catalog # 83008-208
Supplier: Optimize
Specifications
- Description:OPTI-LYNX™ In-line guard holder kit (OPTI-LYNX™ guard cartridge - ordered separately)
- Environmentally Preferable:
- Cat. No.:83008-208
- Supplier no.:11-02738-AA
Specifications
About this item
The OPTI-LYNX™ Guard Column is a high-performance, cartridge-based guard system, suitable for use with any 10 to 32 analytical column.
- For use with OPTI-LYNX™ Guard column cartridges
- Quarter-turn quick-connect design rated to 6,000 psi
- Low Volume/ Low Dispersion interface for volume critical regions
- Repeat zero-dead-volume connection to any column brand
- Hardware is only plumbed once
- Hand-tight - no tools required
OPTI-LYNX combines effective column protection with the convenience of quick-connect operation .Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX utilizes low dispersion component interfaces to keep extra column volume to a minimum: connection paths have a total internal volume of less than 2 μL. You get quick-connect convenience and low dispersion, all in one package.
Delivery: Includes: Bayonet, holder cap, holder tube and fittings.