ContactUsLinkComponent
OPTI-LYNX™ Direct-Connect Trap/Pre-Column Filter Holder Kit
OPTI-LYNX™ Direct-Connect Trap/Pre-Column Filter Holder Kit
Catalog # 83008-168
Supplier:  Optimize
undefined
OPTI-LYNX™ Direct-Connect Trap/Pre-Column Filter Holder Kit
Catalog # 83008-168
Supplier:  Optimize
Supplier Number:  11-02226-AA
Specifications
  • Description:
    OPTI-LYNX™ Direct-connect trap/pre-column filter holder kit
  • Environmentally Preferable:
  • Cat. No.:
    83008-168
  • Supplier no.:
    11-02226-AA
Specifications
About this item
The OPTI-LYNX™ Direct-Connect Trap/Pre-Column Holder System comes complete with the hardware you need to quick-connect to any 10 to 32 valve/port for trapping or analytical column for pre-column filtration.
  • For use with OPTI-LYNX™ Traps and Filters
  • Quarter-turn quick-connect design rated to 6000 psi
  • Low Volume/ Low Dispersion interface for volume critical regions
  • True zero-dead-volume connection to any column brand or 10 to 32 injection valve
  • Hardware is only plumbed once
  • Hand-tight - no tools required
With Opti-Lynx, accessing and changing the insert takes only a quarter turn. Replace traps, filters or change configurations in seconds, without tools. OPTI-LYNX™ traps and filters (ordered separately) come in several bed volume/ diameter/ porosity combinations, allowing you to find your own balance between capacity and volume.
Delivery: Includes: Universal port adapter, holder cap, holder tube and fittings.