ContactUsLinkComponent
ReptiFauna Palm Leaves
ReptiFauna Palm Leaves
Catalog # 470352-314
Supplier:  SEGREST FARMS
undefined
ReptiFauna Palm Leaves
Catalog # 470352-314
Supplier:  SEGREST FARMS
Supplier Number:  963-0117-9
Specifications
  • Description:
    Palm leaves
  • Cat. No.:
    470352-314
Specifications
About this item
Natural decor for high impact and big benefits for your animals.
  • Recreate the rainforest floor to make your pet feel at home
  • Encourages natural behavior of your animals such as burrowing and foraging
  • Long lasting leaves help create a visually dynamic habitat
  • Helps retain soil moisture and ambient humidity
  • Breaks down into nutrients for isopods and plants for a complete bioactive habitat
The addition of palm leaves helps recreate a natural rainforest floor environment for your animal. The curled shape and structure of the leaf provide a variety of hiding places for your isopods and springtails, while and encourage their natural foraging behavior of your pet. Palm Leaves help support a bioactive ecosystem while your isopods and springtails benefit from the breakdown of Palm Leaves as a food source.
Packaging: Leaves are safe to use right out of the package. Simply select the desired amount of leaves and place them into your terrarium. Leaf litter can be scattered on top of the substrate or mixed in for the desired effect.