To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Infliximab
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Size:40 μg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
- Storage Temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2...8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
- Immunogen:Infliximab
- Cat. No.:77007-558
- Packaging:Vial
Infliximab with the brand name of Remicade, is a chimeric monoclonal IgG1 antibody. It binds to TNF-alpha, which is a key proinflammatory cytokine involved in chronic inflammatory diseases. Infliximab functions as TNF-alpha blocker to disrupt the interaction of TNF-alpha with its receptors. GenScript Anti-Infliximab Antibody, pAb, Rabbit is produced from sera of a rabbit immunized with Infliximab.
- High specificity
- High affinity
- High stability
GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for alpha-tubulin and beta-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.
Type: Primary
Antigen: Infliximab
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: