Order Entry
United States
ContactUsLinkComponent
 
Anti-AAV2 Mouse Monoclonal Antibody [clone: 12C10]
 :  Genscript
 
Anti-AAV2 Mouse Monoclonal Antibody [clone: 12C10]
 :  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 77007-556
  • 77007-488
  • 77007-484
  • Antigen Name:
  • AAV2
  • AAV2
  • AAV2
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • 12C10
  • 12C10
  • 12C10
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG2c kappa
  • IgG2c kappa
  • IgG2c kappa
  • Western Blot:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • AAV2
  • AAV2
  • AAV2
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Concentration:
  • Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Cross Adsorption:
  • No
  • No
  • No
  • Storage Buffer:
  • Lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Storage Temperature:
  • The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles

 

Frequently Bought Together