

Ethanol solution 20%, in single-use containers, HyClone™ for bioprocessing
Supplier: Cytiva
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
Flash Pt:30 - 38°C (closed cup)
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Storage:Room Temperature as defined by regional Pharmacopeia (15-30°C in US; 15-25°C in EU)
- Endotoxin Concentration:≤ 1 EU/mL
- Certifications/Compliance:Manufactured in a ISO9001 and ISO13485 certified facility
- Shelf life:12 Months
- For use with:Chromatography resins, IC systems, ÄKTA hardware systems
- Appearance:Clear, colorless solution
- Sterility:No Growth
Specification Test Results
Frequently Bought Together