Order Entry
United States
Orders LinkContactUsLinkComponent
PTS Panels® CHOL + GLU Test Strips

PTS Panels® CHOL + GLU Test Strips

PTS Panels® CHOL + GLU test strips, 20...30 °C (68...86 °F)
Catalog # 77020-346
Supplier:   PTS Diagnostics
Supplier #:  1765
PTS Panels® CHOL + GLU Test Strips
Catalog # 77020-346
Supplier:  PTS Diagnostics
CAS Number:  
PTS Panels® CHOL + GLU Test Strips
PTS Panels® CHOL + GLU Test Strips

PTS Panels® CHOL + GLU test strips, 20...30 °C (68...86 °F)

CAS Number:  
Price
$186.40
Log in for availability
Each
Product Details & Documents

PTS Panels CHOL+GLU test strips provide lab-quality total cholesterol and glucose results on-site in about 45 seconds from a single fingerstick.

  • Measures total cholesterol and glucose
  • 25 test strips per vial
  • Room temperature storage

PTS Panels CHOL+GLU test strips are for professional use with CardioChek PA and CardioChek Plus analyzers to measure total cholesterol and glucose in venous whole blood or capillary fingerstick blood.

This test system will provide numeric results in the following ranges:
Cholesterol: 100 to 400 mg/dl (2.59 to 10.36 mmol/L)
Glucose: 20 to 600 mg/dl (1.11 to 33.3 mmol/L)

Specifications

Specifications

  • Description:
    PTS Panels® CHOL + GLU test strips
  • Storage temperature:
    20...30 °C (68...86 °F)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.