ContactUsLinkComponent
CardioChek PA Blood Sample Analyzer
CardioChek PA Blood Sample Analyzer
  77020-338
undefined
CardioChek PA Blood Sample Analyzer
  77020-338
 :  1708
  • Description:
    CardioChek PA blood sample analyzer
  • Operating Temperature Range:
    20...30 °C (68...86 °F)
  • Storage Temperature Range:
    10...40 °C (50...104 °F)
  • Cat. No.:
    77020-338
The CardioChek PA analyzer achieves accuracy similar to that of a clinical laboratory, but only requires a small (40 μl) blood sample from a fingerstick.
  • Compact, battery-operated
  • Meets NCEP guidelines for accuracy and precision
  • Room temperature test strip storage
  • CLIA Waved
Full lipid panel results in as little as 90 seconds. Single blood chemistry in as little as 45 seconds.
 : CLIA-waived.