ContactUsLinkComponent
3M™ Hot Melt Tapered Aluminum Extension Tip, 9785
3M™ Hot Melt Tapered Aluminum Extension Tip, 9785
Catalog # 76250-804
Supplier:  3M
3M™ Hot Melt Tapered Aluminum Extension Tip, 9785
Catalog # 76250-804
Supplier:  3M
Supplier Number:  7000028621
Specifications
  • Description:
    Hot Melt Tapered Aluminum Extension Tip 9785
  • Dimensions:
    0.070"
  • Cat. No.:
    76250-804
Specifications
About this item
3M™ Hot Melt Applicator Tips offer versatility when dispensing hot melt adhesives. Uniquely designed for our 3M™ Hot Melt Applicators, these tips can help maximize the use of our hot melt adhesives by improving productivity, optimizing precision, and reducing waste in your assembly process.
  • Threaded interiors provides easy installation
  • Metal construction withstands high temperature adhesives
  • Improves sightline and maximum control