ContactUsLinkComponent
3M™ Scotch-Brite™ Multi-Finishing Wheel
3M™ Scotch-Brite™ Multi-Finishing Wheel
Catalog # 76246-234
Supplier:  3M
3M™ Scotch-Brite™ Multi-Finishing Wheel
Catalog # 76246-234
Supplier:  3M
Supplier Number:  7000136471
Specifications
  • Ø:
    6"
  • Description:
    Multi-Finishing Wheel
  • Cat. No.:
    76246-234
  • Grade:
    Fine
  • Abrasive material:
    Silicon carbide
Specifications
About this item
Scotch-Brite™ multi-finishing wheel is a convolute constructed wheel that provides cleaning and final finishing for stainless steel, aluminum, brass, copper and other metals.
  • Silicon Carbide Abrasive
  • Fine grade
  • Runs cool and resists loading due to its open web construction