Orders LinkContactUsLinkComponent
Globe Glass™ Filter Flasks
Globe Glass™ Filter Flasks
Supplier:   Globe Scientific
Globe Glass™ Filter Flasks
Supplier:  Globe Scientific
Certificates
Globe Glass™ Filter Flasks
Globe Glass™ Filter Flasks
8 Options Available Below
Product Details & Documents
Globe Glass™ filter flasks meet ASTM E1406 specifications.
  • Made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass
  • GlassGuard™ packaging system protects against transit damage
  • Side tubulation connects to 7-8 mm flexible tubing
  • Dual metric scale graduations
  • Durable white enamel printing
  • Large white marking area
Thick, heavy-duty walls provide the mechanical strength necessary for filtering operations. Flasks have side tubulation for connection to 7-8 mm flexible tubing. Globe Glass filter flasks are designed with reinforced, beaded rims for strength and resistance to chipping.
Certifications: Meet ASTM E1406, Type II, Class I Specifications.
Documents
Product Family Options
Product Information
  • Capacity
    Material
    Stopper no.
    Diameter ×H
    Graduation Interval
    Graduation Range
    Availability
    Price
  • 76683-704
    Globe Glass™ filter flasks with rim
    Globe Glass™ filter flasks with rim
    25 ml
    Borosilicate glass 3.3
    3
    40×75 mm
    5 ml
    10 - 25 ml
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $182.32
    $182.32
    New Customer Promotional Price
    Box of 6
       $182.32
    Supplier Number: 8450025
  • 76683-746
    Globe Glass™ filter flasks with rim
    Globe Glass™ filter flasks with rim
    50 ml
    Borosilicate glass 3.3
    4
    51×83 mm
    10 ml
    20 - 50 ml
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $187.56
    $187.56
    New Customer Promotional Price
    Box of 6
       $187.56
    Supplier Number: 8450050
  • 76683-738
    Globe Glass™ filter flasks with rim
    Globe Glass™ filter flasks with rim
    125 ml
    Borosilicate glass 3.3
    5
    71×113 mm
    25 ml
    50 - 125 ml
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $191.79
    $191.79
    New Customer Promotional Price
    Box of 6
       $191.79
    Supplier Number: 8450125
  • 76683-728
    Globe Glass™ filter flasks with rim
    Globe Glass™ filter flasks with rim
    250 ml
    Borosilicate glass 3.3
    6
    84×158 mm
    25 ml
    50 - 250 ml
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $213.95
    $213.95
    New Customer Promotional Price
    Box of 6
       $213.95
    Supplier Number: 8450250
  • 76683-668
    Globe Glass™ filter flasks with rim
    Globe Glass™ filter flasks with rim
    500 ml
    Borosilicate glass 3.3
    7
    106×188 mm
    50 ml
    100 - 500 ml
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $273.99
    $273.99
    New Customer Promotional Price
    Box of 6
       $273.99
    Supplier Number: 8450500
  • 76683-742
    Globe Glass™ filter flasks with rim
    Globe Glass™ filter flasks with rim
    1000 ml
    Borosilicate glass 3.3
    8
    135×242 mm
    50 ml
    250 - 1000 ml
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $465.79
    $465.79
    New Customer Promotional Price
    Box of 6
       $465.79
    Supplier Number: 8451000
  • 76683-744
    Globe Glass™ filter flasks with rim
    Globe Glass™ filter flasks with rim
    2000 ml
    Borosilicate glass 3.3
    9
    165×288 mm
    100 ml
    600 - 2000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $103.26
    $103.26
    New Customer Promotional Price
    Each
       $103.26
    Supplier Number: 8452000
  • 76683-736
    Globe Glass™ filter flasks with rim
    Globe Glass™ filter flasks with rim
    4000 ml
    Borosilicate glass 3.3
    12
    205×360 mm
    250 ml
    1000 - 4000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $208.66
    $208.66
    New Customer Promotional Price
    Each
       $208.66
    Supplier Number: 8454000