ContactUsLinkComponent
CardioChek Plus Analyzer
CardioChek Plus Analyzer
Catalog # 76994-108
Supplier:  PTS Diagnostics
undefined
CardioChek Plus Analyzer
Catalog # 76994-108
Supplier:  PTS Diagnostics
Supplier Number:  2700
Specifications
  • Description:
    CardioChek Plus analyzer
  • Storage Temperature Range:
    10...40 °C (50...104 °F)
  • Cat. No.:
    76994-108
Specifications
About this item
The CardioChek Plus analyzer achieves accuracy similar to that of a clinical laboratory, but only requires a small (40 μl) blood sample from a fingerstick.
  • Full lipid panel results in as little as 90 seconds
  • Meets NCEP guidelines for accuracy and precision
  • Portable, lightweight, and battery-powered
  • Refrigerated storage of test strips is not required
  • CLIA Waved
Providers spend less time testing and more time counseling patients face-to-face during the same office visit for that 'teachable moment'. The CardioChek test system meets NCEP guidelines for accuracy and precision, CRMLN-certfified, and reimbursable in most instances.
Certifications: CLIA-waived, FDA-cleared.