Specifications
- Cat. No.:76994-104
- Storage temperature:−30...−10 °C (−22…+14 °F)
Specifications
About this item
A1CNow+ Controls are intended for the quality control of PTS Diagnostics' A1CNow+ HbA1c tests. The controls are supplied in two levels as a frozen, ready to use material that requires no reconstitution.
- Ready-to-use control materials
- Two-level control kit
- Dropper vials
The PTS Diagnostics A1CNow+ test system is fast, easy, and accurate. It provides A1C results in 5 minutes which are 99% laboratory accurate*. These A1C tests are for use in a doctor’s office. Providing real-time results outside of the laboratory, A1CNow+ is a hand-held, portable monitor which allows A1C testing in every exam room. Real-time A1C results allow for timely decisions on therapy changes when needed. The product requires no maintenance. Office staff no longer have to prepare and refrigerate lab samples. Obtaining A1C results during the patient office visit eliminates follow-up phone calls to discuss results and possible therapeutic changes. Our A1CNow® System is CLIA waived and certified by the National Glycohemoglobin Standardization Program (NGSP).
Once the control is thawed and opened, it may be stored refrigerated at 2 to 8 °C (36 to 46 °F) for 8 months or at room temperature at 18 to 25 °C (64 to 77 °F) for 7 days. The control remains stable until the expiration date printed on the label.