Order Entry
United States
ContactUsLinkComponent
PTS Panels® Lipid + eGLU Smart Bundle
PTS Panels® Lipid + eGLU Smart Bundle
Catalog # 76994-110
Supplier:  PTS Diagnostics
CAS Number:  
undefined
PTS Panels® Lipid + eGLU Smart Bundle
Catalog # 76994-110
Supplier:  PTS Diagnostics
Supplier Number:  2729
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Cat. No.:
    76994-110
  • Storage temperature:
    20...30 °C (68...86 °F)

Specifications

About this item

PTS Panels Lipid + eGLU Smart Bundle allows you to get accurate lipid and blood glucose results in as little as 90 seconds, from one fingerstick.

  • For use with the CardioChek Plus analyzer
  • 15-count lipid test strips vial and one 15-count electrochemical glucose (eGLU) test strips vial
  • Accurate results in as little as 90 seconds

PTS Panels Lipid Panel+eGLU test strips are for professional use exclusively with the CardioChek Plus analyzer to measure total cholesterol, high density lipoprotein (HDL) cholesterol, Triglycerides and glucose in venous whole blood or capillary fingerstick blood.

Cholesterol measurements are used in the diagnosis and treatments of disorders involving excess cholesterol in the blood and lipid and lipoprotein metabolism disorders. Lipoprotein measurements are used in the diagnosis and treatment of lipid disorders (such as diabetes mellitus), atherosclerosis, and various liver and renal diseases. Glucose measurements are used in the diagnosis and treatment of carbohydrate metabolism disorders including diabetes mellitus, neonatal hypoglycemia, and idiopathic hypoglycemia, and of pancreatic islet carcinoma.

This test system will display numeric results in the following ranges:
Cholesterol: 100 to 400 mg/dl (2.59 to 10.36 mmol/l)
Hdl Cholesterol: 20 to 120 mg/dl (0.52 to 3.11 mmol/l)
Triglycerides: 50 to 500 mg/dl (0.57 to 5.65 mmol/l)
Glucose: 40 to 600 mg/dl (2.22 to 33.3 mmol/l)

Certifications: CLIA-waived, FDA-cleared.