Anti-HCV Human Recombinant Antibody [clone: RV36]
This antibody has been tested to neutralize VSV Pseudovirus HCV E1E2 (H77 strain) (Catalog P2R77).
- Catalog No:
- 77007-408
- 77007-444
- Antigen Symbol:
- HCV
- HCV
- Antigen Name:
- Structural envelope E2
- Structural envelope E2
- Antigen Synonyms:
- E
- E
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV36
- RV36
- Reactivity:
- Human
- Human
- Host:
- Human
- Human
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Modification:
- Recombinant monoclonal antibody
- Recombinant monoclonal antibody
- Molecular Weight:
- 150 kDa
- 150 kDa
- Purification:
- Purified by Protein A chromatography
- Purified by Protein A chromatography
- Concentration:
- 1 mg/ml by refractometry
- 1 mg/ml by refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping Temperature:
- Ice
- Ice
Frequently Bought Together