Order Entry
United States
ContactUsLinkComponent
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide 98%
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide 98%
Catalog # 76981-294
Supplier:  Ambeed
CAS Number:  
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide 98%
Catalog # 76981-294
Supplier:  Ambeed
Supplier Number:  A733995-10G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    10 g
  • Cat. No.:
    76981-294

Specifications

About this item

 :
 :275.73 g/mol
 :GHS07
CAS Number:41859-57-8
Formula:C₁₅H₁₄ClNO₂
MDL Number:MFCD00218513