Orders LinkContactUsLinkComponent
BD MAX™ LBC Sample Buffer Tubes
BD MAX™ LBC sample buffer tubes, pack of 48
  76906-232
 :   BD
 :  443923
BD MAX™ LBC Sample Buffer Tubes
  76906-232
 :  BD
BD MAX™ LBC Sample Buffer Tubes
BD MAX™ LBC Sample Buffer Tubes
BD MAX™ LBC sample buffer tubes, pack of 48
$158.46
Pack of 48
BD MAX™ liquid based cytology (LBC) sample buffer tubes.
  • Liquid based cytology specimens in ThinPrep® PreservCyt® solution
  • No re-capping step by lab technician after vortex
  • Post-testing, a sample buffer tube with a pierced cap may also be stored upright for up to 4 days at 2 to 30 °C
  • Description:
    BD MAX™ LBC sample buffer tubes, pack of 48