ContactUsLinkComponent
BD MAX™ Urine Transport Kit
BD MAX™ Urine Transport Kit
Catalog # 76906-234
Supplier:  BD
undefined
BD MAX™ Urine Transport Kit
Catalog # 76906-234
Supplier:  BD
Supplier Number:  443924
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BD MAX™ urine transport kit, pack of 96
  • Cat. No.:
    76906-234
Specifications
About this item
BD MAX™ urine transport kit used for collection of male and female urine.
  • Male and female urine collection device, introduction of new pierceable caps
  • Eliminates re-capping step by lab technician and helps to reduce waste (1 cap/sample)
  • Post-testing, a sample buffer tube with a pierced cap may also be stored upright for up to 4 days at 2 to 30 °C
  • Transport stability at 2 to 30 ⁰C vs. UVE specimen collection kit (21
  • days vs. 5 days)
  • Elimination of Pre-warm step for urine sample