ContactUsLinkComponent
BD MAX™ Swab Collection Kit
BD MAX™ Swab Collection Kit
  76906-236
 :  BD
undefined
BD MAX™ Swab Collection Kit
  76906-236
 :  BD
 :  443925
  • Description:
    BD MAX™ swab collection kit, pack of 100
  • Cat. No.:
    76906-236
BD MAX™swab collection kit.
  • Multiple BD assays from one swab
  • Eliminates re-capping step by lab technician and helps to reduce waste (1 cap/sample)
  • Post-testing, a sample buffer tube with a pierced cap may also be stored upright
  • for up to 4 days at 2 to 30 °C
  • Up to 4x longer transport stability vs. UVE specimen collection Kit
  • for BD CTGCTV2 for BD MAX™ (21 days vs. 5 days)
  • Elimination of Pre-warm step for swab samples