BD MAX™ CDIFF Kit
BD MAX™ CDIFF test kit, 24
Catalog # 76906-212
Supplier: BD
Supplier #: 443418
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The BD MAX™ CDIFF test enables accurate and rapid identification of Clostridium difficile infections for supporting infection control practice.
- Clostridium difficile
- Molecular
The BD MAX™ CDIFF test identifies patients with CDI, including hypervirulent strains, by targeting the toxin B gene (tcdB), which is specific for toxigenic C. difficile.
Specifications
Specifications
- Description:BD MAX™ CDIFF test kit
- No. of tests:24