ContactUsLinkComponent
BD MAX™ GBS Kit
BD MAX™ GBS Kit
  76906-208
 :  BD
 :  
undefined
BD MAX™ GBS Kit
  76906-208
 :  BD
 :  441772
 :  
  • Description:
    BD MAX™ GBS test kit
  • Cat. No.:
    76906-208
  • No. of tests:
    24
The BD MAX™ GBS assay provides timely and accurate Group B Streptococcus screening results to help reduce the risk of GBS disease in newborn babies
  • Group B Streptococcus
  • Detects GBS DNA in Lim Brothcultures from antepartum women
  • Molecular
The BD MAX™ GBS assay, run on the fully-automated,easy-to-use BD MAX™ System, detects GBS DNA in Lim Broth cultures from antepartum women.4 The BD accommodates on-demand and batch workflows to meet MAX™ System your laboratory’s needs.